missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ II Reverse Transcriptase

Produktkod. p-4926554
missing translation for 'orderingAttributeHoverText'
No. of Reactions:
10 Reactions
200 Reactions
50 Reactions
Förpackningsstorlek
2ku
Each
Pack of 4
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 10635003

missing translation for 'mfr': Invitrogen™ 18064022

Please to purchase this item. Need a web account? Register with us today!

Edge
Denna artikel kan inte returneras. Se returpolicy

Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT.

Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT. Mutations in the RNase H domain of the enzyme eliminate degradation of the RNA during first-strand cDNA synthesis, which results in higher yields of full-length cDNA.

SuperScript reverse transcriptases are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.

Features of SuperScript II Reverse Transcriptase include:
• High cDNA yields and more full-length product
• Generation of cDNA up to 12.3 kb
• First-strand cDNA synthesis at higher temperatures than conventional MMLV RT

Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase (Cat. No. 18090010), features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.

Bestellinformation

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Spezifikation

Concentration 200 U/μL
Content And Storage

• SuperScript II Reverse Transcriptase, 10 μL (1 x 2,000 units at 200 U/μL)
• 5X First-strand buffer
• 100 mM DTT

Store at –20°C.

Format Stand-alone Enzyme
GC-Rich PCR Performance High
Reaction Speed 50 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 2,000 Units
Reverse Transcriptase SuperScript II
Ribonuclease H Activity Reduced
Shipping Condition Wet Ice
Final Product Type First-Strand cDNA
No. of Reactions 10 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 12.3 kb
Starting Material RNA
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt