Learn More
Invitrogen™ SuperScript™ VILO™ cDNA Synthesis Kit
Description
Includes
5X VILO Reaction Mix (200μL) and a 10X SuperScript III Enzyme Blend (100μL), which contains: SuperScript III RT, RNaseOUT Recombinant Ribonuclease Inhibitor, and a proprietary helper protein
The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes. The enzyme blend contains SuperScript III Reverse Transcriptase (RT), a genetically engineered MMLV RT that has reduced RNase H activity and improved thermostability for highly efficient cDNA synthesis.
The SuperScript VILO cDNA Synthesis Kit can be used to synthesize cDNA from for a wide range of input RNA amounts. This kit provides the high temperature capability of SuperScript III Reverse Transcriptase in an optimized format for generating first-strand cDNA for use in RT–qPCR.
Notes
• For superior first-strand cDNA synthesis in two-step RT-qPCR, try SuperScript IV VILO Master Mix (Cat. No. 11756050) or SuperScript IV VILO Master Mix with ezDNase Enzyme (Cat. No. 11766050).
• This kit is included with EXPRESS Two-Step RT–qPCR kits. Reagents are provided for 50 or 250 20-μL cDNA synthesis reactions.
Informations sur la commande
Shipping Condition: Dry ice
Spécification
Spécification
| Content And Storage | • 5X VILO Reaction Mix, 200 μL |
| Detection Method | qPCR |
| Format | Kit |
| GC-Rich PCR Performance | High |
| Reaction Speed | 60 min. |
| Technique | Reverse Transcription |
| Optimal Reaction Temperature | 42°C |
| Quantity | 50 reactions |
| Reverse Transcriptase | SuperScript III |
| Ribonuclease H Activity | Reduced |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.