missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO

Displays bright fluorescence when bound to RNA and low background in gels. Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO is ideal for use with either formaldehyde/agarose or polyacrylamide gels.

Brand:  Invitrogen™ S7568

 View more versions of this product

Product Code. 10463252

  • 980.00 € / 1mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SYBR Green II RNA gel stain is one of the most sensitive dyes known for detecting RNA in electrophoretic gels using laser scanners or standard UV transilluminators. It is significantly more sensitive than ethidium bromide, the most commonly used stain for detecting nucleic acids in gels.

Available in three convenient packaging options:

  • 500μL (stains 50 minigels)
  • 1mL (stains 100 minigels)
  • 20 × 50μL (each stains 5 minigels)—Avoids waste when processing fewer minigels

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Fluorescence
RNA Gel Stain
Room Temperature
In-Gel Detection
SYBR Green II
1 mL
RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.