missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Safe DNA Gel Stain in 0.5X TBE
GREENER_CHOICE

Developed specifically for reduced mutagenicity – to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels

Brand:  Invitrogen™ S33101

378.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11550676

  • 630.00 € / 4 L
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SYBR Safe stain is less mutagenic than ethidium bromide and offers better sensitivity than ethidium bromide. Supplied as a premixed solution that can be used like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe stain has fluorescence excitation maxima at ∽280 and ∽502nm, and an emission maximum at ∽530nm. SYBR Safe DNA gel stain is also available as 4L (S-33101) of ready-to-use solution. The SYBR Safe DNA Gel Stain Starter Kit (S-33110, sold separately) includes 1L of SYBR Safe DNA gel stain and one photographic filter (S-37100).

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Supplied as a ready-to-use solution in 0.5X TBE

Store at room temperature in the original container.
Fluorescence
DNA Gel Stain
4 L
DNA
In-Gel Detection
SYBR Safe
Less hazardous
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.