missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE
GREENER_CHOICE

Developed specifically for reduced mutagenicity to be less hazardous. Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE is safer and more sensitive than ethidium bromide for staining DNA in agarose or acrylamide gels. The premixed solution is available in 1L and 4L units.

Brand:  Invitrogen™ S33112

382.80 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11560676

  • 638.00 € / 4 L
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SYBR Safe™ stain is not only less mutagenic than ethidium bromide, but SYBR Safe™ stain′s detection sensitivity is better than that of ethidium bromide. SYBR Safe™ stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe™ stain has fluorescence excitation maxima at ∼280 and ∼502nm, and an emission maximum at ∼530nm.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Supplied as a ready-to-use solution in 1X TAE

Store at room temperature in the original container.
Fluorescence
DNA Gel Stain
4 L
DNA
In-Gel Detection
SYBR Safe
Less hazardous
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.