missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Safe Imager™ Viewing Glasses
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Each
missing translation for 'unitSize'
Each
Description
Includes
I pair of Safe Imager™ viewing glasses
The Safe Imager viewing glasses allow visualization of gel bands without use of the Safe Imager amber filter to facilitate band excision.
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Content And Storage | I pair of Safe Imager™ viewing glasses |
| For Use With (Equipment) | Safe Imager™ |
| Product Line | SYBR Safe |
| Type | Viewing Glass |
| Quantity | 1 Each |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction