missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T4 DNA Polymerase

DNA polymerase with 3'-exodeoxyribonuclease activity, but not 5'a3' exodeoxyribonuclease activity

Brand:  Invitrogen™ 18005025

 View more versions of this product

Product Code. 10043202

  • 790.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: T4 DNA Polymerase is supplied with a vial of 5X T4 DNA Polymerase buffer [165mM Tris acetate (pH 7.9), 330mM sodium acetate, 50mM magnesium acetate, 5mM DTT].
This item is not returnable. View return policy

Description

Description

  • Applications: Labeling double-stranded linear DNA by replacement synthesis, Oligonucleotide-directed, site-specific mutagenesis, 3' end-labeling of double-stranded DNA, Polishing both 5' or 3 overhangs to make blunt ends
  • Source: Purified from E. coli expressing the T4 DNA Polymerase gene on a plasmid
  • Performance and Quality Testing: Single- and double-stranded endodeoxyribonuclease and phosphatase assays; exodeoxyribonuclease and polymerase activities tested
  • Unit Definition: One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using DNase I-nicked DNA as template primer
  • Unit Reaction Conditions: 50mM glycine-NaOH (pH 8.8), 16.6mM (NH4)2SO4, 6mM MgCl2, 6.5μM EDTA, 10mM 2-mercaptoethanol, 0.165mg/mL BSA, 1.6mg/mL DNase I-nicked salmon testes DNA, 0.33mM dCTP, 0.33mM dATP, 0.33mM dGTP, 0.33mM dTTP, 76nM [3H]dTTP, and enzyme in 0.1mL for 30 min. at 37°C
  • A T4 DNA Polymerase Technical Bulletin is available

ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics and Non-Coding RNA Research, Restriction Enzyme Cloning

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

T4 DNA Polymerase is supplied with a vial of 5X T4 DNA Polymerase buffer [165 mM Tris acetate (pH 7.9), 330 mM sodium acetate, 50 mM magnesium acetate, 5 mM DTT]. Store at -20°C.
T4 DNA Polymerase
250 U
No
DNA Polymerase
3' - 5'
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2508677 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18005025
Lot Number2614573 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number18005025
Lot Number2769029 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number18005025
Lot Number2964772 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number18005025
Lot Number3002608 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number18005025
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ T4 DNA Polymerase > 250 units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.