Learn More
Invitrogen™ Total Exosome Isolation Kit (from plasma)
The Total Exosome Isolation Kit (from plasma) consists of two parts: Total Exosome Isolation Reagent (from plasma) and Proteinase K (20 mg/mL).
Brand: Invitrogen™ 4484450
442.70 EUR valid until 2025-12-31
Use promo code "24338" to get your promotional price.
Includes: 6mL bottle of Total Exosome Precipitation Reagent (from plasma) and 1.25mL bottle of Proteinase K for Total Exosome Isolation (from plasma) (20 mg/mL).
Description
The Total Exosome Isolation Kit (from plasma) consists of two parts: Total Exosome Isolation Reagent (from plasma) and Proteinase K (20 mg/mL). The Total Exosome Isolation Reagent enables simple and reliable concentration of intact exosomes from plasma samples with a protocol that is scalable depending on your sample size. Proteinase K is provided separately for optional upfront plasma treatment and is used to eliminate all plasma protein and thus maximize the purity of the exosome preparation.
- Maximize recovery of intact exosomes for any type of downstream application
- Easily isolate exosomes using our simple and reliable protocol
- Avoid time-consuming ultra-centrifugation
- Maximize flexibility—scale up or down depending on the sample size
Specifications
• 6 mL bottle of Total Exosome Precipitation Reagent (from plasma). Store at 2–8°C. • 1.25 mL bottle of Proteinase K for Total Exosome Isolation (from plasma) (20 mg/mL). Store at -5 to -30°C. |
|
Exosomes | |
6 mL |
Total Exosome Isolation | |