missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ DEPC-Treated Water
GREENER_CHOICE

Autoclaved to remove the DEPC and 0.1-μm membrane-filtered.

50.50 € - 81.75 €

Specifications

Grade Molecular Biology
Treatment(s) DEPC-Treated
Purification Method Autoclaved, Membrane-Filtered
Product Line UltraPure™
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11588636
View Documents
Invitrogen™
10813012
4 x 1.25 mL
81.75 €
Pack of 4
In Stock
to see stock.
Add to basket
 
11528616
View Documents
Invitrogen™
750024
4 x 100 mL
50.50 €
400mL
In Stock
to see stock.
Edge
Add to basket
 
10636503
GREENER_CHOICE
View Documents
Invitrogen™
750023
1 L
74.00 €
1 L
In Stock
to see stock.
Add to basket
 
Description

Description

UltraPure™ DEPC-treated Water is suitable for use with RNA. It is prepared by incubating with 0.1% diethylpyrocarbonate (DEPC), and is then autoclaved to remove the DEPC. Sterile filtered.

Performance and Quality Testing: No DNase or RNase activity detected.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Molecular Biology
DEPC-Treated
Autoclaved, Membrane-Filtered
UltraPure™
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store in freezer (-5 to -30°C).

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.