Learn More
Invitrogen™ UltraPure™ Ethidium Bromide, 0.625 mg/mL
Fluorescent dye used to detect nucleic acids in agarose gels and cesium chloride (CsCl) gradient
Brand:  Invitrogen™ A25645
Product Code. 15361527
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
1 x 1 mL ethidium bromide dropper (0.625 mg/mL)
Store at room temperature.
Gel Electrophoresis
Ethidium Bromide
Room Temperature
In-Gel Detection
Ethidium bromide
UltraPure™
DNA & RNA