missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ Glycerol
UltraPure Glycerol is suitable for the concentration and storage of enzymes at low temperatures. A 50% (v/v) solution will not freeze at -20°C.
Brand: Invitrogen™ 15514011
87.20 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- UltraPure Glycerol is suitable for the concentration and storage of enzymes at low temperatures
- A 50% (v/v) solution will not freeze at -20°C
- Since it has a density of 1.26 g/mL, glycerol frequently is used as a component in electrophoresis loading buffers
- UltraPure Glycerol gradients can be used to purify proteins, bacteriophage, or organelles
- Redistilled UltraPure Glycerol is ≥ 99% pure
- No DNase, RNase, or protease activity detected
- Store at room temperature
Order Info
Shipping Condition: Room temperature
Specifications
Glycerol | |
UltraPure™ | |
500 mL | |
Liquid |
Store at room temperature. | |
Molecular Biology Grade | |
Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction