missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)

UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl.

245.00 € - 417.00 €

Specifications

Chemical Name or Material Phenols
pH 8.05
Product Line UltraPure™
Purity Molecular Biology Grade
Shipping Condition Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11518756
View Documents
Invitrogen™
15593031
100 mL
245.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11528756
View Documents
Invitrogen™
15593049
400 mL
417.00 €
400mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl
  • When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase
  • UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives
  • It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
  • The appearance of the solution is evaluated at room temperature
  • No RNase or DNase activity is detected
  • Store in the refrigerator at 2°C to 8°C

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Phenols
UltraPure™
Room Temperature
8.05
Molecular Biology Grade
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • UltraPure Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H341-Suspected of causing genetic defects.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Phenol, Chloroform,1-Butanol, 3-methyl-

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.