missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ SSC, 20X
GREENER_CHOICE

Product Code. 10515203
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 10515203

Brand: Invitrogen™ 15557044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in nucleic acid hybridizations and blot transfer applications

UltraPure 20X SSC is a solution formulated and used in concentrations ranging from 0.2X to 20X, depending on the application.

  • Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™ Box
  • Sterile-filtered UltraPure 20X SSC contains 3.0 M NaCl and 0.3 M sodium citrate, at pH 7.0
  • Performance and Quality Testing: No DNase or RNase activity detected

DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Concentration 20 X
Product Type SSC
Form Liquid
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Quantity 1 L
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.