missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ TBE Buffer, 10X

Product Code. 11508986
Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
Each
This item is not returnable. View return policy

Product Code. 11508986

Brand: Invitrogen™ 15581028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Compatible with agarose gels

UltraPure 10X TBE Buffer is a sterile-filtered solution of 1M Tris, 0.9 M boric acid, and 0.01M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis.

  • UltraPure 10X TBE Buffer is available in 1 L plastic bottle/10 L Cubitainer™ box
  • Performance and quality testing: No DNase, RNase, or protease activity detected. Sustainable packaging

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Running Buffer
Concentration 10 X
Content And Storage Store at room temperature.
Format Bottle
Gel Compatibility Polyacrylamide Gels, Agarose Gels
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Product Type TBE Buffer
Quantity 10 L
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.