Learn More
Invitrogen™ UltraPure™ Agarose
Easy-pour spout reduces spills and contamination
Brand: Invitrogen™ 16500100
162.67 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose is ideal for resolving DNA and RNA fragments from 100 bp to >30 kb
- Ideal for analysis and recovery of DNA and RNA for routine applications
- Strong gel structure allows for better handling and less breakage
- Can be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
- Improved packaging
- Performance and quality testing
Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Contents: 1 pouch containing 100 g of UltraPure™ Agarose Storage: 15°C to 30°C |
|
100 to >30000 bp | |
Gel Electrophoresis | |
Sustainable packaging | |
UltraPure™ | |
Room Temperature |
Agarose | |
Powder | |
Agarose Gels | |
Standard Melting Point | |
100 g |
For Research Use Only. Not for use in diagnostic procedures.