Learn More
Invitrogen™ Yeast Deletion Heterozygous Essential, Complete
Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project
Brand:  Invitrogen™ 95401.H5R3
Product Code. 10277124
  • 6190.00 € / Each
Explore more special offers
Description
Description
Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yeast Deletion Heterozygous Essential, Complete
Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418)
16 Plates
Yeast deletion library created by a collaboration of 8 North American and 8 European laboratories composing the Saccharomyces Genome Deletion Project
Complete Set
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Yeast Deletion Heterozygous Essential, Complete > Heterozygous Diploid Essential Complete Set