missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast Nitrogen Base

Base medium for preparation of minimal and synthetic defined yeast media

Brand:  Invitrogen™ Q30009

Product Code. 11573667

  • 726.00 € / 500g
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: Each pouch contains reagents to prepare 500mL of 10X YNB for Pichia media or 1000mL of 10X YNB for S. cerevisiae media.
This item is not returnable. View return policy

Description

Description

  • YNB contains ammonium sulfate but does not contain amino acids
  • Autoclavable

Cloning, Transformation

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Each bottle contains 500 g of reagent. Use 67 g to prepare 500 mL of 10X YNB for Pichia media or 1,000 mL of 10X YNB for S. cerevisiae media.

Store at room temperature. Guaranteed stable for 12 months when properly stored.
YNB
Microbiology Media
Liquid Medium
500 g/Bottle
Room Temperature
Powder
Autoclave
P. pastoris, S. cerevisiae
Powder Reagents
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.