missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast tRNA

Product Code. p-4921718
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
This item is not returnable. View return policy

Product Code. 11508736

Brand: Invitrogen™ 15401011

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Supplied in lyophilized form

Yeast tRNA is suitable for use as a carrier in nucleic acid purification and precipitation procedures.

Array CGH, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Microarray Analysis

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Yeast tRNA
Content And Storage Store in freezer (-5 to -30°C).
Reagent Type Yeast tRNA
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
For Use With (Application) Carrier for Nucleic Acid Precipitation
Quantity 25 mg
Form Lyophilized Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.