missing translation for 'onlineSavingsMsg'
Learn More

Iodic acid, 99.6%, Thermo Scientific Chemicals

Product Code. 10509413
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O

Spezifikation

Color White
Melting Point 110.0°C
Density 4.6200g/mL
Heavy Metals (as Pb) 10ppm max.
Packaging Glass bottle
Residue after Ignition 0.02% max.
Specific Gravity 4.62
Assay Percent Range 99.6%
Quantity 500 g
Linear Formula HIO3
Physical Form Crystalline Powder
Merck Index 15, 5056
Solubility Information Solubility in water: 2690g/l (20°C). Other solubilities: soluble in nitric acid, soluble in diluted alcohol, insoluble in absolute alcohol, ether and, chloroform
Formula Weight 175.91
Percent Purity 99.6%
Grade ACS Reagent
Chemical Name or Material Iodic acid, For analysis ACS
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'