Learn More
Iodic acid, 99%, Thermo Scientific Chemicals
Description
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.
Solubility
Soluble in water and methanol.
Notes
Light sensitive. Incompatible with reducing agents, alcohols and organic materials.
Specifications
Specifications
Melting Point | 110°C (decomposition) |
Density | 4.629 |
Odor | Odorless |
Sensitivity | Light sensitive |
Quantity | 250 g |
UN Number | UN3085 |
Merck Index | 14,5012 |
Synonym | unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 175.91 |
Show More |
Safety and Handling
- Iodic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 A
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Iodic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.