missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodide Iodate concentrate, For 1L standard solution, 1/128 M I2 (1/64N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 Pc. |
UN Number | NONH for all modes of transport |
Beilstein | 3587194 |
IUPAC Name | potassium iodide |
Molecular Weight (g/mol) | 166.00 |
Packaging | Ampoule |
Chemical Name or Material | Iodide Iodate concentrate |
Safety and Handling
Product Identifier
- Iodide Iodate concentrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : potassium hydroxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'