missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodide solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
77.00 € - 114.00 €
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12942135
|
Fisher Chemical
A 022 250 |
250 mL | Bottle |
77.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12952135
|
Fisher Chemical
A 022 500 |
500 mL | Bottle |
114.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
I− |
Specifications
1g/L | |
I− | |
Liquid |
Single-component Standard Solution | |
H2O | |
Iodide |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title