missing translation for 'onlineSavingsMsg'
Learn More

Iodide Standard For Ion Chromatography, SPEX CertiPrep™

Product Code. 11985074
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
I-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Identificatori chimici

CAS 7681-82-5, 7732-18-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifica

Packaging Triple-leached LDPE bottle
Quantity 500 mL
CAS Max % 99.9, 0.1
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition (by Analyte or Components) 1000mg/L
Analyte or Component Names Sodium iodide,distilled water,conductivity or of similar purity
Solvent or Matrix H2O
Chemical Name or Material Iodide Standard, For Ion Chromatography

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'