missing translation for 'onlineSavingsMsg'
Learn More
Iodine, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals Product Code.: 10123510

Iodine, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals

Product Code. 10123510
1 L, Glass bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10123510

Brand: Thermo Scientific Acros 124220010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2, 7681-11-0, 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Brown
Physical Form Liquid
Quantity 1 L
Linear Formula I2
Fieser 01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153
Merck Index 15, 5057
Formula Weight 253.81
Concentration or Composition (by Analyte or Components) 0.0998 to 0.1002N (20°C)
Packaging Glass bottle
Density 1.0200g/mL
Chemical Name or Material Iodine, 0.1N Standard Solution
Show More Show Less
compliance-icons
Product Identifier
  • Iodine, 0.1N standard solution
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.