missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 99.5+%, Extra Pure, SLR, Fisher Chemical™

50.65 € - 142.00 €

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10694722
View Documents Promotion Details
Fisher Chemical
I/0450/48
100 g
50.65 €
100g
Please to purchase this item. Need a web account? Register with us today!
10500921
View Documents Promotion Details
Fisher Chemical
I/0450/50
250 g
83.65 €
250g
Please to purchase this item. Need a web account? Register with us today!
10694912
View Documents Promotion Details
Fisher Chemical
I/0450/53
500 g
142.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7553-56-2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
II
I2
MFCD00011355 MFCD00164163
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606

Specifications

185°C
7553-56-2
5.1
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
II
253.81
CHEBI:17606
Amber glass bottle
Vapor Density: 8.8
113°C
Gray
Solid
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
molecular iodine
807
253.81g/mol
0.41 hPa 25
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iodine

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications