missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Reagent Grade, ≥99.8% (titration), Honeywell™

Product Code. 15697900
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.8% (titration)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Boiling Point 184°C (literature)
Melting Point 113°C (literature)
CAS Max % 100.0000%
Quantity 100 g
Linear Formula Fe2(SO4)3 · xH2O
UN Number UN3495
Beilstein 3587194
Formula Weight 253.81
Percent Purity >99.8%
Grade Reagent
Packaging Glass Bottle
Density 4.930 g/cm3; Vapor Density: 9 (vs air)
Chemical Name or Material Iodine, Reagent Grade, ≥99.8% (titration)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'