missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, ACS Reagent, Honeywell Fluka™
ACS Reagent, solid,≥99.8%
42.62 € - 1534.64 €
Chemical Identifiers
CAS | 7553-56-2 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15661780
|
Honeywell-Fluka
207772-5G |
5 g | N/A | ||||||
15621780
|
Honeywell-Fluka
207772-100G |
100 g |
117.77 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15651780
|
Honeywell-Fluka
207772-500G |
500 g |
380.46 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15631780
|
Honeywell-Fluka
207772-1KG |
1 kg |
707.01 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15641780
|
Honeywell-Fluka
207772-2.5KG |
2.5 kg |
1534.64 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15671780
|
Honeywell-Fluka
207772-6X500G |
6 x 500 g | N/A | ||||||
Description
Honeywell ACS solvents for general laboratory use are:
- a value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested tmeet ACS general use specifications
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 |
Specifications
7553-56-2 | |
I2 | |
3495 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
≥99.8% | |
0.31mmHg (25°C), 1mmHg (38.7°C) | |
Iodine |
Solid | |
MFCD00011355 MFCD00164163 | |
3587194 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
diiodine | |
807 | |
253.81g/mol | |
ACS Reagent | |
Vapor Density: 9 (vs air) |
Safety and Handling
P261-P273-P280-P305 + P351 + P338-P314
H312 + H332-H315-H319-H335-H372-H400
missing translation for 'einecsNumber' : 231-442-4
missing translation for 'rtecsNumber' : NN1575000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Iodine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title