missing translation for 'onlineSavingsMsg'
Learn More

Iodine Bromide Solution, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11494689
Click to view available options
Quantity:
1 L
Packaging:
Tamper-evident
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured according to European Pharmacopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Iodine Bromide Solution, EP Grade, Reagecon™ is a ready-to-use reagent for use in manual or automatic titrations. It is prepared by adding 20g IBr to 1L glacial acetic acid.

Iodine monobromide is an interhalogen compound that is used in some types of iodometry as a source of I+.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use reagent eliminates the time and labor of preparing precise reagents and volumetric solutions

Specifications

CAS 7789-33-5, 64-19-7
Color Orange-Red
Physical Form Liquid
Quantity 1 L
Molecular Formula BrI
MDL Number MFCD00011353
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
Solubility Information Freely soluble in water, in alcohol and in glacial acetic acid
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
SMILES BrI
IUPAC Name iodobromane
Molecular Weight (g/mol) 206.81
PubChem CID 82238
Formula Weight 318.3g/mol
Grade Eu. Ph. Chapter 4(4.1.1)
Concentration or Composition (by Analyte or Components) 20g IBr in 1L glacial acetic acid
Odor Vinegar
Packaging Tamper-evident
Chemical Name or Material Iodine monobromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • IODINE BROMIDE SOLUTION
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Acetic Acid, IodineBromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'