missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Certified AR for Analysis, Fisher Chemical™
53.00 € - 1615.00 €
Chemical Identifiers
CAS | 7553-56-2 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10439750
|
Fisher Chemical
I/0500/48 |
100 g |
53.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10152000
|
Fisher Chemical
I/0500/50 |
250 g |
125.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10162000
|
Fisher Chemical
I/0500/53 |
500 g |
1615.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 |
Specifications
185°C | |
7553-56-2 | |
5.1 | |
I2 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
Amber glass bottle | |
Vapor Density: 8.8 |
113°C | |
Gray | |
Solid | |
MFCD00011355 MFCD00164163 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
diiodine | |
807 | |
253.81g/mol | |
0.41 hPa 25 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Iodine
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P273-Avoid release to the environment.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title