missing translation for 'onlineSavingsMsg'
Learn More

Iodine concentrate, For 1L standard solution, 0.05 M I2 (0.1 N), Honeywell Fluka™

Product Code. 15685200
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

for 1L standard solution, 0.05 M I2 (0.1 N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0, 7553-56-2, 7732-18-5, 7732-18-5

Specifications

CAS Min % ≥50.0000%, ≥20.0000%
CAS Max % <70.0000%, <25.0000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
Molecular Formula IK
Linear Formula I2
MDL Number MFCD00011355
UN Number NONH for all modes of transport
Beilstein 3587194
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
PubChem CID 4875
ChEBI CHEBI:8346
Formula Weight 253.81
Packaging Ampoule
Chemical Name or Material Iodine concentrate, For 1L standard solution, 0.05 M I2 (0.1 N)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodine concentrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iodine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'