Learn More
Iodine monochloride, 1M solution in dichloromethane, Thermo Scientific Chemicals
Specifications
Chemical Name or Material | Iodine monochloride |
---|---|
Name Note | 1, 0M solution in dichloromethane |
CAS | 7790-99-0, 75-09-2 |
Color | Brown to Orange |
Physical Form | Solution |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Iodine monochloride | |
7790-99-0, 75-09-2 | |
Solution | |
ICl | |
01,502; 08,261; 10,212; 11,268 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
0.95 to 1.05M | |
1.42 |
1, 0M solution in dichloromethane | |
Brown to Orange | |
ClI | |
MFCD00011354 | |
15, 5060 | |
Solubility in water: insoluble | |
ClI | |
162.35 | |
162.35 | |
Glass bottle | |
1.4200g/mL |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
R
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-236-7
- Iodine monochloride, 1,0M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only