Learn More
Iodine monochloride, 98%, Thermo Scientific Chemicals
65.70 € - 224.00 €
Chemical Identifiers
CAS | 7790-99-0 |
---|---|
Molecular Formula | ClI |
Molecular Weight (g/mol) | 162.35 |
MDL Number | MFCD00011354 |
InChI Key | QZRGKCOWNLSUDK-UHFFFAOYSA-N |
Synonym | iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french |
PubChem CID | 24640 |
IUPAC Name | iodochlorane |
SMILES | ClI |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10206880
|
Thermo Scientific Acros
196961000 |
100 g | Glass bottle |
65.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10771661
|
Thermo Scientific Acros
196965000 |
500 g | Glass bottle |
224.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7790-99-0 | |
162.35 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
24640 | |
ClI |
ClI | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
iodochlorane |
Specifications
97.0°C | |
7790-99-0 | |
Fuming Liquid | |
ClI | |
MFCD00011354 | |
15, 5060 | |
Solubility in water: insoluble. Other solubilities: decomposes, soluble in ethanol, ether and cs2 | |
ClI | |
162.35 | |
162.36 | |
3.2 | |
Iodine monochloride, 98% |
26.0°C | |
Red to Brown | |
97.5% min. (Iodometry) | |
ICl | |
01,502; 08,261; 10,212; 11,268 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
98% | |
3.2000g/mL |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-236-7
missing translation for 'rtecsNumber' : NN1650000
missing translation for 'tsca' : TSCA
- Iodine monochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H300-Fatal if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only