missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine monochloride, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Boiling Point | 97.0°C |
Melting Point | 26.0°C |
Color | Brown-Red |
Physical Form | Fuming Liquid |
Quantity | 500 g |
Linear Formula | ICl |
Fieser | 01,502; 08,261; 10,212; 11,268 |
Merck Index | 15, 5060 |
Solubility Information | Solubility in water: insoluble. Other solubilities: decomposes, soluble in ethanol, ether and cs2 |
Formula Weight | 162.36 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction