missing translation for 'onlineSavingsMsg'
Learn More

Iodine monochloride, ACS, Thermo Scientific Chemicals

Moisture and Light Sensitive

52.66 € - 631.14 €

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11408490
SDS View Documents
Thermo Scientific Alfa Aesar
044348.14
25 g
52.66 €
25g
Please to purchase this item. Need a web account? Register with us today!
11498480
SDS View Documents
Thermo Scientific Alfa Aesar
044348.22
100 g
148.74 €
100g
Please to purchase this item. Need a web account? Register with us today!
11488480
SDS View Documents
Thermo Scientific Alfa Aesar
044348.36
500 g
631.14 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7790-99-0
162.35
QZRGKCOWNLSUDK-UHFFFAOYSA-N
24640
ClI
ClI
MFCD00011354
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
iodochlorane

Specifications

96°C to 98°C
7790-99-0
Crystalline, Liquid
MFCD00011354
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodochlorane
24640
ACS
Red-black crystalline/liquid
Iodine monochloride, For analysis ACS
25°C to 27°C
Black-Red
ClI
3902972
Soluble in water,alcohol,ether,carbon disulfide.
ClI
162.35
162.36
Light and moisture sensitive
3.182
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300-H311-H314-H335-H373

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 232-236-7

missing translation for 'rtecsNumber' : NN1650000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Iodine monochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Iodine monochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.