Learn More
Iodine monochloride, approx. 0.22N soln. in glacial acetic acid, Thermo Scientific Chemicals
For Determination of Iodine Absorption Number of Fats
87.33 € - 133.67 €
Chemical Identifiers
CAS | 7790-99-0 |
---|---|
Molecular Formula | ClI |
Molecular Weight (g/mol) | 162.35 |
MDL Number | MFCD00011354 |
InChI Key | QZRGKCOWNLSUDK-UHFFFAOYSA-N |
Synonym | iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french |
PubChem CID | 24640 |
IUPAC Name | iodochlorane |
SMILES | ClI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11336129
|
Thermo Scientific Alfa Aesar
042647.AP |
500 mL |
87.33 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345367
|
Thermo Scientific Alfa Aesar
042647.K2 |
1 L |
133.67 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For determination of iodine absorption number of fatsIodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Moisture and Light SensitiveChemical Identifiers
7790-99-0 | |
162.35 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
24640 | |
ClI |
ClI | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
iodochlorane |
Specifications
7790-99-0 | |
ClI | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
485°C (905°F) (acetic acid) | |
Liquid |
Liquid | |
MFCD00011354 | |
Miscible with water,alcohol,ether and carbon disulfide. | |
ClI | |
162.35 | |
162.36 | |
Light and moisture sensitive | |
Iodine monochloride |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8, Packing Group: II
missing translation for 'einecsNumber' : 232-236-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only