missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 99+%, pure

Product Code. 10574463 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 10574463

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Color Gray
Physical Form Spheres
Quantity 100 g
Linear Formula I2
Merck Index 15, 5057
IUPAC Name diiodine
Formula Weight 253.81
Percent Purity ≥99.5%
Grade Extra Pure
Packaging Glass bottle
Vapor Pressure 0.41hPa at 25°C
Density 8.8 (Air = 1.0)
Chemical Name or Material Iodine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iodine
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.