missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Reagent ACS, 99.8%, Thermo Scientific Chemicals

27.34 € - 681.39 €

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10493551
View Documents
Thermo Scientific Acros
423820050
5 g Glass Bottle
27.34 €
5g
Please to purchase this item. Need a web account? Register with us today!
10065940
View Documents
Thermo Scientific Acros
423825000
500 g Glass Bottle
285.42 €
500g
Please to purchase this item. Need a web account? Register with us today!
10609293
View Documents
Thermo Scientific Acros
423820025
2.5 kg Glass bottle
681.39 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

7553-56-2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
II
I2
MFCD00011355 MFCD00164163
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606

Specifications

185°C
7553-56-2
Prills
I2
MFCD00011355 MFCD00164163
01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
II
253.81
CHEBI:17606
≥99.80%
0.41hPa at 25°C
Iodine, 99.8%
113°C
Gray
99.8+%
I2
3495
15, 5057
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
807
253.81
ACS Reagent
8.8 (Air = 1.0)
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove to fresh and keep confortable for several minutes.

Warning

missing translation for 'einecsNumber' : 231-442-4

missing translation for 'rtecsNumber' : NN1575000

missing translation for 'tsca' : TSCA

Product Identifier
  • Iodine

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.