missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Reagent Grade, beads, 1-3 mm, 99.7% trace metals basis, Honeywell™

Product Code. 15642810
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, beads, 1-3 mm, 99.7% trace metals basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Boiling Point 184°C (literature)
Melting Point 113°C (literature)
CAS Max % 100.0000%
Physical Form Beads
Quantity 250 g
Linear Formula NaH2PO2 · xH2O
UN Number UN3495
Beilstein 3587194
Formula Weight 253.81
Percent Purity ≥99.7%
Grade Reagent
Packaging Glass Bottle
Density 4.930 g/cm3
Chemical Name or Material Iodine, Reagent Grade, beads, 1-3 mm, 99.7% trace metals basis
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'