Learn More
Iodine Solution, 0.005M, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Quantity | 100 mL |
Concentration or Composition (by Analyte or Components) | 0.005mol/L |
Safety and Handling
Product Identifier
- Iodine 0.05M Volumetric solution
- Warning
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : iodine