missing translation for 'onlineSavingsMsg'
Learn More

Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N), Honeywell Fluka™

Product Code. 15604800
Click to view available options
Quantity:
2.5 L
5 L
4 x 2.5 L
6 x 1 L

Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2, 7681-11-0, 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II

Specifications

CAS Min % ≥90.0000%, ≥1.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <5.0000%, <5.0000%
Quantity 6 x 1 L
Linear Formula I2
Beilstein 3587194
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
IUPAC Name diiodine
Grade Reagent Ph. Eur.
Concentration or Composition (by Analyte or Components) 0.05 M I2 (0.1N)
Packaging Glass Bottle
Chemical Name or Material Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N)
Show More Show Less
compliance-icons
Product Identifier
  • Iodine solution
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iodine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'