missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Solution, Volumetric, 0.025M I2 (0.05N), Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Quantity | 100 mL |
Beilstein | 3587194 |
Formula Weight | 253.81g/mol |
Packaging | Glass Bottle |
Appearance | Liquid |
Density | 1.03g/mL at 25°C |
Chemical Name or Material | Iodine solution, Volumetric, 0.025M I2 (0.05N) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction