missing translation for 'onlineSavingsMsg'
Learn More

Iodine trichloride, 97%, Thermo Scientific Chemicals

35.48 € - 320.60 €

Chemical Identifiers

CAS 865-44-1
Molecular Formula Cl3I
Molecular Weight (g/mol) 233.254
MDL Number MFCD00036291
InChI Key PAWIVBWALDNUJP-UHFFFAOYSA-N
Synonym iodine trichloride, iodine chloride icl3, unii-1e5kq66trq, 1e5kq66trq, jodtrichlorid, trichloro-, e3-iodane, trichloro-$l^ 3-iodane, ksc450m5f, iodine trichloride, saj first grade
PubChem CID 70076
IUPAC Name trichloro-$l^{3}-iodane
SMILES ClI(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15482418
SDS View Documents
Thermo Scientific Alfa Aesar
H56005.06
5 g
35.48 €
5g
Please to purchase this item. Need a web account? Register with us today!
15492418
SDS View Documents
Thermo Scientific Alfa Aesar
H56005.14
25 g
107.54 €
25g
Please to purchase this item. Need a web account? Register with us today!
15402428
SDS View Documents
Thermo Scientific Alfa Aesar
H56005.22
100 g
320.60 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

Solubility
Soluble with decomposition.

Notes
Light Sensitive. Store at -20°C.
Specifications

Chemical Identifiers

865-44-1
233.254
PAWIVBWALDNUJP-UHFFFAOYSA-N
70076
ClI(Cl)Cl
Cl3I
MFCD00036291
iodine trichloride, iodine chloride icl3, unii-1e5kq66trq, 1e5kq66trq, jodtrichlorid, trichloro-, e3-iodane, trichloro-$l^ 3-iodane, ksc450m5f, iodine trichloride, saj first grade
trichloro-$l^{3}-iodane

Specifications

865-44-1
ICl3
UN3085
iodine trichloride, iodine chloride icl3, unii-1e5kq66trq, 1e5kq66trq, jodtrichlorid, trichloro-, e3-iodane, trichloro-$l^ 3-iodane, ksc450m5f, iodine trichloride, saj first grade
PAWIVBWALDNUJP-UHFFFAOYSA-N
trichloro-$l^{3}-iodane
70076
97%
Iodine trichloride
Cl3I
MFCD00036291
14,5020
Soluble with decomposition.
ClI(Cl)Cl
233.254
233.26
Light sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 212-739-8

missing translation for 'tsca' : Yes

Recommended Storage : Store at -20°C

Product Identifier
  • Iodine trichloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Iodine chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.