Learn More
Iodine(V) oxide, 98%, Thermo Scientific Chemicals
An oxidizing agent to remove CO from gas
76.08 € - 213.06 €
Chemical Identifiers
CAS | 12029-98-0 |
---|---|
Molecular Formula | I2O5 |
Molecular Weight (g/mol) | 333.804 |
MDL Number | MFCD00011356 |
InChI Key | BIZCJSDBWZTASZ-UHFFFAOYSA-N |
Synonym | iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide |
PubChem CID | 159402 |
ChEBI | CHEBI:29914 |
IUPAC Name | iodyl iodate |
SMILES | O=I(=O)OI(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11365019
|
Thermo Scientific Alfa Aesar
087682.14 |
25 g |
76.08 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375019
|
Thermo Scientific Alfa Aesar
087682.22 |
100 g |
213.06 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iodine (V) oxide is used as an oxidizing agent to remove carbon monoxide from gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12029-98-0 | |
333.804 | |
BIZCJSDBWZTASZ-UHFFFAOYSA-N | |
159402 | |
iodyl iodate |
I2O5 | |
MFCD00011356 | |
iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide | |
CHEBI:29914 | |
O=I(=O)OI(=O)=O |
Specifications
275°C to 350°C (decomposition) | |
White to Cream | |
Crystalline | |
MFCD00011356 | |
14,5019 | |
Soluble in water and nitric acid; Insoluble in absolute alcohol,chloroform,ethanol,ether and carbon disulfide. | |
O=I(=O)OI(=O)=O | |
333.804 | |
CHEBI:29914 | |
98% | |
5.08 g/mL |
12029-98-0 | |
1.1 | |
I2O5 | |
UN3085 | |
iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide | |
BIZCJSDBWZTASZ-UHFFFAOYSA-N | |
iodyl iodate | |
159402 | |
333.81 | |
Hygroscopic; Light sensitive | |
Iodine (V) oxide |
Safety and Handling
GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: III
missing translation for 'einecsNumber' : 234-740-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.