missing translation for 'onlineSavingsMsg'
Learn More
Iodoacetic acid, 97+%, Thermo Scientific Chemicals Product Code.: 11489583

Iodoacetic acid, 97+%, Thermo Scientific Chemicals

Product Code. 11489583
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11489583

Brand: Thermo Scientific Alfa Aesar A12410.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.

Solubility
Soluble in water, alcohol and ethanol. Slightly soluble in ether and chloroform.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-69-7
Molecular Formula C2H3IO2
Molecular Weight (g/mol) 185.948
MDL Number MFCD00002685
InChI Key JDNTWHVOXJZDSN-UHFFFAOYSA-N
Synonym iodoacetic acid, acetic acid, iodo, iodoacetate, monoiodoacetic acid, monoiodoacetate, monoiodine acetate, iodoaceticacid, iodoethanoic acid, acetic acid, 2-iodo, kyselina jodoctova
PubChem CID 5240
ChEBI CHEBI:74571
IUPAC Name 2-iodoacetic acid
SMILES C(C(=O)O)I

Specifications

Melting Point 79°C to 82°C
Boiling Point 208°C (decomposition)
Odor Pungent
Quantity 25 g
UN Number UN2923
Beilstein 1739079
Sensitivity Light sensitive
Merck Index 14,5025
Solubility Information Soluble in water,alcohol and ethanol. Slightly soluble in ether and chloroform.
Formula Weight 185.95
Percent Purity ≥97%
Chemical Name or Material Iodoacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodoacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iodoacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoacetic acid, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.