missing translation for 'onlineSavingsMsg'
Learn More
Iodoacetic acid, 97%, Thermo Scientific Chemicals Product Code.: 10449230

Iodoacetic acid, 97%, Thermo Scientific Chemicals

Product Code. 10449230
100 g, Glass bottle
Click to view available options
Quantity:
25 g
50 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10449230

Brand: Thermo Scientific Acros 122281000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-69-7
Molecular Formula C2H3IO2
Molecular Weight (g/mol) 185.95
MDL Number MFCD00002685
InChI Key JDNTWHVOXJZDSN-UHFFFAOYSA-N
Synonym iodoacetic acid, acetic acid, iodo, iodoacetate, monoiodoacetic acid, monoiodoacetate, monoiodine acetate, iodoaceticacid, iodoethanoic acid, acetic acid, 2-iodo, kyselina jodoctova
PubChem CID 5240
ChEBI CHEBI:74571
IUPAC Name 2-iodoacetic acid
SMILES C(C(=O)O)I

Specifications

Melting Point 79.0°C to 83.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ICH2CO2H
Quantity 100 g
Beilstein 02, 222
Merck Index 15, 5068
Solubility Information Solubility in water: 600g/L (20 c). Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 185.95
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Iodoacetic acid, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463407 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number122280500,122281000,122280250
Lot NumberA0436287 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number122280500,122281000,122280250
Lot NumberA0449166 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number122280500,122281000,122280250
Lot NumberA0401484 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number122280500,122281000,122280250
Lot NumberA0415190 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number122280500,122281000,122280250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoacetic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.