missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetic acid, sodium salt, 99%, Thermo Scientific Chemicals

Product Code. 10245940
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-53-3
Molecular Formula C2H2INaO2
Molecular Weight (g/mol) 207.93
MDL Number MFCD00002686
InChI Key AGDSCTQQXMDDCV-UHFFFAOYSA-M
Synonym sodium iodoacetate, iodoacetic acid sodium salt, acetic acid, iodo-, sodium salt, sodium 2-iodoacetate, sodium monoiodoacetate, iodoacetate, sodium salt, sodium iodacetate, iodoacetic acid, sodium salt, unii-07814f2qri, ccris 9232
PubChem CID 5239
IUPAC Name sodium;2-iodoacetate
SMILES C(C(=O)[O-])I.[Na+]

Specifications

Melting Point 208.0°C to 210.0°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ICH2CO2Na
Quantity 100 g
Beilstein 02, 222
Fieser 13,116
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 207.93
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Iodoacetic acid, sodium salt
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodoacetic acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iodoacetic acid, sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'