missing translation for 'onlineSavingsMsg'
Learn More
Iodoacetonitrile, 95%, Thermo Scientific Chemicals Product Code.: 10081260

Iodoacetonitrile, 95%, Thermo Scientific Chemicals

Product Code. 10081260
5 mL, Glass bottle
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10081260

Brand: Thermo Scientific Acros 122290050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-75-9
Molecular Formula C2H2IN
Molecular Weight (g/mol) 166.95
MDL Number MFCD00001886
InChI Key VODKOOOHHCAWFR-UHFFFAOYSA-N
Synonym iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
PubChem CID 69356
IUPAC Name 2-iodoacetonitrile
SMILES C(C#N)I

Specifications

Color Brown to Red
Density 2.3000g/mL
Boiling Point 182°C to 184°C (720.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ICH2CN
Refractive Index 1.5740 to 1.578
Quantity 5 mL
Beilstein 02, 223
Specific Gravity 2.3
Formula Weight 166.95
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Iodoacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • Iodoacetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoacetonitrile, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.