missing translation for 'onlineSavingsMsg'
Learn More
Iodoacetonitrile, 97%, Thermo Scientific Chemicals Product Code.: 11332256

Iodoacetonitrile, 97%, Thermo Scientific Chemicals

Product Code. 11332256
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11332256

Brand: Thermo Scientific Alfa Aesar L03634.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

Solubility
Soluble in benzene, alcohol, acetone, ether. Slightly soluble in water. Insoluble in hexane.

Notes
Light Sensitive. Store at 4°C. Incompatible with reducing agents, light and bases. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-75-9
Molecular Formula C2H2IN
Molecular Weight (g/mol) 166.949
MDL Number MFCD00001886
InChI Key VODKOOOHHCAWFR-UHFFFAOYSA-N
Synonym iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
PubChem CID 69356
IUPAC Name 2-iodoacetonitrile
SMILES C(C#N)I

Specifications

Density 2.306
Boiling Point 70°C to 72°C (10 mmHg)
Flash Point 86°C (186°F)
Linear Formula ICH2CN
Refractive Index 1.574
Quantity 10 g
UN Number UN2922
Beilstein 1734628
Sensitivity Light sensitive
Solubility Information Soluble in benzene,alcohol,acetone,ether. Slightly soluble in water. Insoluble in hexane.
Formula Weight 166.95
Percent Purity 97%
Chemical Name or Material Iodoacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • Iodoacetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Iodoacetonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoacetonitrile, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.