missing translation for 'onlineSavingsMsg'
Learn More

Iodobenzene diacetate, 98%, Thermo Scientific Chemicals

Product Code. 11994911
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3240-34-4
Molecular Formula C10H11IO4
Molecular Weight (g/mol) 322.09
InChI Key ZBIKORITPGTTGI-UHFFFAOYSA-N
Synonym iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo
PubChem CID 76724
IUPAC Name [acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
SMILES CC(=O)OI(C1=CC=CC=C1)OC(=O)C

Specifications

Melting Point 161.0°C to 165.0°C
Color Colorless to Yellow
Loss on Drying 1% max. (60°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Linear Formula C6H5I(O2CCH3)2
Quantity 500 g
Fieser 01,508; 03,166; 04,266; 10,214; 13,242
Solubility Information Solubility in water: insoluble
Formula Weight 322.09
Percent Purity 98%
Physical Form Solution
Chemical Name or Material Iodobenzene diacetate
Show More Show Less
compliance-icons
Product Identifier
  • Iodobenzene diacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'