Learn More
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -108.0°C |
Color | Colorless to Yellow or Pink |
Density | 1.9500g/mL |
Boiling Point | 69.0°C to 73.0°C |
Flash Point | 62°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.5100 to 1.5130 |
Quantity | 100 g |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0449610 | Certificate of Analysis | 20/03/2024 | 332125000,332121000,332120025 |
A0435949 | Certificate of Analysis | 06/11/2023 | 332125000,332121000,332120025 |
A0446924 | Certificate of Analysis | 24/10/2023 | 332125000,332121000,332120025 |
A0397052 | Certificate of Analysis | 14/12/2022 | 332125000,332121000,332120025 |
A0409934 | Certificate of Analysis | 13/12/2022 | 332125000,332121000,332120025 |
Safety and Handling


- Iodoethane, stabilized
- Danger
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.